Learn More
Medchemexpress LLC (Rac)-Idroxioleic acid (2-hydroxyoleic acid) | 56472-29-8 | 99.9% | 1 MG
SDP
Supplier:  Medchemexpress LLC HY12946710MM/1ML
Encompass_Preferred
(Rac)-Idroxioleic acid (2-Hydroxyoleic acid) is a synthetic oleic acid (OA) derivative that binds to the plasma membrane and alters lipid organization. It demonstrates anti-tumor effects, impairing Jurkat cell growth in a time- and concentration-dependent manner in vitro with an IC50 of approximately 40 μM, and inducing apoptosis. In vivo, it significantly inhibits tumor growth in nude mice bearing Jurkat cell xenografts.
  • Synthetic oleic acid (OA) derivative.
  • Binds to the plasma membrane and alters lipid organization.
  • Exhibits anti-tumor effects.
  • Impairs Jurkat cell growth (IC50 ~40 μM) and induces apoptosis in vitro.
  • Significantly inhibits tumor growth in Jurkat cell xenograft models in vivo.
Catalog No. 50-003-40144