Learn More
Medchemexpress LLC (Rac)-Idroxioleic acid (2-Hydroxyoleic acid) | 56472-29-8 | 99.9% | 1 MG
SDP
Supplier:  Medchemexpress LLC HY1294671MG
Encompass_Preferred
(Rac)-Idroxioleic acid (2-Hydroxyoleic acid) is a synthetic oleic acid (OA) derivative that binds to the plasma membrane and alters lipid organization. It has an anti-tumor effect. It is for research use only.
  • Binds to the plasma membrane and alters lipid organization
  • Has an anti-tumor effect
  • Impairs Jurkat cell growth in a time- and concentration-dependent manner
  • Induces a marked and concentration-dependent increase in the proteolytic cleavage of PARP
  • Inhibits tumor growth in nude mice
Catalog No. 50-003-41229