Learn More
Medchemexpress LLC (Rac)-Idroxioleic acid (2-hydroxyoleic acid) | 56472-29-8 | 99.9% | 100 MG

Supplier: Medchemexpress LLC HY129467100MG
(Rac)-Idroxioleic acid (2-hydroxyoleic acid) is a synthetic oleic acid (OA) derivative that binds to the plasma membrane and alters lipid organization. It exhibits anti-tumor effects.
- Binds to the plasma membrane and alters lipid organization
- Impairs Jurkat cell growth in a time and concentration dependent manner in vitro
- Induces proteolytic cleavage of PARP, a molecular marker of apoptosis
- Significantly inhibits tumor growth in nude mice in vivo
- Molecular weight: 298.46
- Formula: C18H34O3
- Appearance: Solid, white to off-white
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.