Learn More
Medchemexpress LLC rac -Isoleucine 500g
SDP
Supplier:  Medchemexpress LLC HYW1005647500G
Encompass_Preferred
(rac)-Isoleucine is the racemic form of Isoleucine Isoleucine is a mixture of the diastereomers L-isoleucine L-alloisoleucine D-isoleucine and D-alloisoleucine L-isoleucine is a branched-chain amino acid that acts as a nitrogen donor and has roles in glucose consumption fatty acid metabolism as well as immune function L-Alloisoleucine is generated by transamination of L-isoleucine D-Isoleucine and D-alloisoleucine are enantiomers of L-isoleucine and L-alloisoleucine respectively[1][2][3]
Catalog No. 50-002-05437