Learn More
Medchemexpress LLC (Rac)-JBJ-04-125-02 acetate | 00-00-0 | MFCD32709390; MFCD16606534 | 98.7% | 543.61 g/mol | C29H26FN5O3S | 5 MG
SDP
Supplier:  Medchemexpress LLC HY135805B5MG
Encompass_Preferred
(Rac)-JBJ-04-125-02 acetate is the racemic acetate salt of a research-grade allosteric EGFR inhibitor. It is a potent, mutant-selective, orally active compound with reported sub-nanomolar biochemical potency against the L858R/T790M EGFR variant and demonstrated single-agent activity in cellular and animal models. Supplied as a white to off-white solid for laboratory research use only.
  • Potent, mutant-selective EGFR inhibition with sub-nanomolar biochemical potency.
  • Demonstrated single-agent activity in both in vitro and in vivo models.
  • Allosteric mode of action complementary to ATP-site inhibitors.
  • Available as a racemic acetate solid suitable for assay and formulation work.
  • Provided with documentation such as COA, HPLC, and NMR upon request.
Catalog No. 50-002-21868