Learn More
Medchemexpress LLC Rac-lanicemine (AZD6765) | 61890-25-3 | MFCD00924222 | 99.6% | 198.27 g/mol | C13H14N2 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY108235B10MM 1ML
Encompass_Preferred
(Rac)-Lanicemine is the racemic form of lanicemine (AZD6765), a low-trapping NMDA receptor channel blocker used in neuroscience research. Supplied as a concentrated solution for in vitro and analytical applications, it is intended for use under appropriate laboratory safety conditions and storage recommendations.
  • Racemic NMDA receptor antagonist for neuropharmacology studies.
  • Supplied as a concentrated solution for direct use in assays.
  • High chemical purity suitable for analytical applications.
  • Stable when stored according to recommended conditions.
  • Provided with product documentation such as datasheet and certificate of analysis.
Catalog No. 50-002-44006