Learn More
Medchemexpress LLC (Rac)-LM11A-31 (dihydrochloride) | 1214672-15-7 | 99.4% | 316.27 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY110155A10MM/1ML
Encompass_Preferred
(Rac)-LM11A-31 dihydrochloride is a significant research chemical that acts as a non-peptide modulator of the p75NTR (neurotrophin receptor p75). It is presented as an isomer and racemate of LM11A-31, demonstrating its role as an orally effective proNGF inhibitor. This compound is primarily utilized in research settings to explore pathways related to neurotrophin receptors.
  • Functions as a non-peptide modulator of p75NTR
  • An isomer and racemate of LM11A-31
  • Acts as an orally effective proNGF inhibitor
  • Primarily for research use
Catalog No. 50-003-44788