Learn More
Medchemexpress LLC (rac)-lomedeucitinib | 2328064-33-9 | 98.1% | 419.47 g/mol | C18H17D3N6O4S | 10 MG

Supplier: Medchemexpress LLC HY160144S210MG
rac-lomedeucitinib is the racemic form of lomedeucitinib provided as a solid research compound for in-vitro and preclinical studies. It is intended for laboratory research use only; consult the SDS for handling, storage, and safety information.
- Racemic small-molecule TYK2 inhibitor.
- Provided as a solid powder suitable for laboratory use.
- High purity: 98.1%.
- Molecular weight: 419.47 g/mol.
- CAS number: 2328064-33-9.
- Available in small pack sizes, including 10 mg.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.