Learn More
Medchemexpress LLC (Rac)-lunresertib | 2719749-28-5 | 99.7% | C18H20N4O2 | 1 ML

Supplier: Medchemexpress LLC HY14581710MM/1ML
(Rac)-lunresertib is a potent membrane-associated tyrosine and threonine-specific cdc2-inhibitory kinase (Myt1) inhibitor. It exhibits an IC50 of less than 10 nM and demonstrates anticancer effects, making it suitable for various research applications.
- Potent Myt1 inhibitor
- Inhibitor with IC50 of less than 10 nM
- Demonstrates anticancer effects
- Molecular weight is 324.38
- Soluble in DMSO at 100 mg/mL
- Appears as a solid, off-white to yellow color
- For research use only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.