Learn More
Medchemexpress LLC (Rac)-LY341495 | 201943-63-7 | >96.5% | C20H19NO5 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY70059A100MG
Encompass_Preferred
(Rac)-LY341495 is an isomer of LY341495 (HY-70059) and can be used as a control compound in experiments. LY341495 is a metabotropic glutamate receptor (mGluR) antagonist.
  • Suitable for use as a control compound in experiments
  • Functions as a metabotropic glutamate receptor (mGluR) antagonist
  • Exhibits IC50s for mGlu2 (21 nM), mGlu3 (14 nM), mGlu1a (7.8 μM), mGlu5a (8.2 μM), mGlu8 (170 nM), mGlu7 (990 nM), and mGlu4 (22 μM) receptors.
Catalog No. 50-004-34788