Learn More
Medchemexpress LLC (Rac)-LY341495 | 96.5% | C20H19NO5 | 10 MM * 1 ML

Supplier: Medchemexpress LLC HY70059A10MM/1ML
(Rac)-LY341495 is an isomer of LY341495 and can be utilized as a control compound in experiments. The parent compound, LY341495, functions as a metabotropic glutamate receptor (mGluR) antagonist, exhibiting IC50 values of 21 nM for mGlu2, 14 nM for mGlu3, 7.8 μM for mGlu1a, 8.2 μM for mGlu5a, 170 nM for mGlu8, 990 nM for mGlu7, and 22 μM for mGlu4 receptors.
- Isomer of LY341495.
- Can be used as a control compound in experiments.
- LY341495 is a metabotropic glutamate receptor (mGluR) antagonist.
- Molecular weight: 353.37
- Appearance: Solid
- Color: White to off-white
- For research use only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.