Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (Rac)-LY341495 | 96.5% | C20H19NO5 | 50 MG
SDP

Supplier:  Medchemexpress LLC HY70059A50MG

Encompass_Preferred

(Rac)-LY341495 is an isomer of LY341495 (HY-70059) that acts as a metabotropic glutamate receptor (mGluR) antagonist. It serves as a control compound in experiments and is relevant in neurological and eye/ear disease research.

  • Used as a control compound in experiments
  • Acts as a metabotropic glutamate receptor antagonist
  • Molecular weight: 353.37
  • Appearance: Solid, white to off-white
  • Solubility in DMSO: 2.5 mg/mL
  • Relevant to GPCR/G protein and neuronal signaling pathways

Catalog No. 50-004-34813


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.