Learn More
Medchemexpress LLC (Rac)-m826 | 2649883-37-2 | 99.8% | 575.70 g·mol⁻¹ | C28H45N7O6 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY155612A10MG
Encompass_Preferred
(Rac)-M826 is the racemic form of a selective and reversible caspase-3 inhibitor used in apoptosis and nervous system disease research. It has an enzymatic IC50 of 0.005 μM, inhibits apoptosis in NT2 cells, and is supplied as a high-purity powder for biochemical and cell-based studies.
  • Potent caspase-3 inhibition (IC50 0.005 μM).
  • Inhibits apoptosis in NT2 cells, supporting cellular research.
  • High purity (>99.7%) suitable for sensitive assays.
  • Supplied as a stable powder for convenient handling and storage.
  • Available in small research-scale quantities.
Catalog No. 50-002-86298