Learn More
Medchemexpress LLC (Rac)-Metanephrine (hydrochloride) | 881-95-8 | 99.8% | 233.69 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYW012206S15MG
Encompass_Preferred
(Rac)-Metanephrine hydrochloride is a metabolite of epinephrine and a substrate of the sulfotransferase SULT1A3. It undergoes sulfonation at its p-hydroxyl group via a reaction catalyzed by SULT1A3. It is an isomer of Metanephrine hydrochloride, and it is applicable to the research of pheochromocytoma.
  • For research use only.
  • Metabolite of epinephrine.
  • Substrate of sulfotransferase SULT1A3.
  • Applicable to the research of pheochromocytoma.
Catalog No. 50-004-53657