Learn More
Medchemexpress LLC (Rac)-Metanephrine (hydrochloride) | 881-95-8 | 99.8% | 233.69 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYW0122065MG
Encompass_Preferred
(Rac)-Metanephrine hydrochloride is an epinephrine metabolite and a sulfotransferase SULT1A3 substrate. It undergoes sulfonation at its p-hydroxyl group via a reaction catalyzed by SULT1A3. This compound is an isomer of Metanephrine (HY-113299) hydrochloride and is used in pheochromocytoma research.
  • Metabolite of epinephrine
  • Substrate for sulfotransferase SULT1A3
  • Undergoes sulfonation at p-hydroxyl group
  • Isomer of Metanephrine hydrochloride
  • Applicable to pheochromocytoma research
  • Inhibits MPP+ uptake into Caki-1 cells and rat hepatocytes
Catalog No. 50-004-52790