Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Rac-MGV354 | 1852495-86-3 | 575.7 | C35H37N5O3 | 1 ML
SDP

Supplier:  Medchemexpress LLC HY11791710MM 1ML

Encompass_Preferred

(Rac)-MGV354 is the racemic form of MGV354, a potent soluble guanylate cyclase (sGC) activator supplied for research use. It exhibits sub-nanomolar to low-nanomolar cellular activity and is intended for in vitro studies.

  • Racemic form of MGV354
  • Target: soluble guanylate cyclase (sGC)
  • Cellular activity: EC50 <0.5 nM (CHO cells), 5 nM (GTM-3E cells)
  • Molecular formula C35H37N5O3; molecular weight 575.7
  • Typical supplied format: 10 mM solution, 1 mL; DMSO stock preparations described
  • Storage: solid at 4°C protected from light; in solution store at -80°C (up to 6 months) or -20°C (up to 1 month)
  • Purity: not specified in the supplier datasheet

Catalog No. 50-002-26611


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.