Learn More
Medchemexpress LLC Rac-MGV354 | 1852495-86-3 | MFCD32664441 | 99.3% | 575.7 | C35H37N5O3 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY117917100MG
Encompass_Preferred
(Rac)-MGV354 is the racemic form of MGV354, a potent soluble guanylate cyclase (sGC) activator used in biochemical and cellular research. It demonstrates sub-nanomolar to low-nanomolar activity in cell assays and is used to study guanylate cyclase-mediated signaling.
  • Potent sGC activation with EC50s of <0.5 nM (CHO) and 5 nM (GTM-3 E).
  • Suitable for biochemical and cellular assays investigating cGMP signaling.
  • Solid, white to off-white form with molecular weight 575.7 and formula C35H37N5O3.
  • Storage: 4°C, protect from light; in solvent: -80°C (6 months), -20°C (1 month).
Catalog No. 50-002-26571