Learn More
Medchemexpress LLC (Rac)-minzasolmin | 1802518-63-3 | 99.9% | 425.59 g/mol | C23H31N5OS | 10MG
SDP
Supplier:  Medchemexpress LLC HY12528710MG
Encompass_Preferred
(Rac)-Minzasolmin is a racemic, brain-penetrant inhibitor of α-synuclein misfolding developed for research into synucleinopathies. It modulates early-stage aggregation, can revert membrane-bound oligomers to monomers, and has shown efficacy in α-synuclein transgenic models relevant to Parkinson's disease and dementia with Lewy bodies.
  • Brain-penetrant small molecule suitable for in vivo and in vitro studies.
  • Inhibits α-synuclein misfolding and early-stage aggregation.
  • Promotes reversion of membrane-bound oligomers to monomers.
  • Demonstrated activity in α-synuclein transgenic models.
  • High purity for research use (≈99.9%).
  • Molecular formula C23H31N5OS; molecular weight 425.59 g/mol.
Catalog No. 50-002-37235