Learn More
Medchemexpress LLC (Rac)-Modipafant | 122956-68-7 | 98.15% | 605.09 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY108908100MG
Encompass_Preferred
(Rac)-Modipafant (UK-74505) is an orally active, selective, long-acting irreversible platelet activating factor receptor (PAFR) antagonist that prevents dengue infection. This product is for research use only.
  • Orally active.
  • Selective.
  • Long-acting irreversible platelet activating factor receptor (PAFR) antagonist.
  • Prevents dengue infection.
  • Exhibits highly selective, time-dependent inhibition of PAF-induced aggregation of rabbit washed platelets.
  • Dose-dependently inhibits zymosan-induced articular hyperalgesia.
Catalog No. 50-003-45368