Learn More
Medchemexpress LLC (Rac)-Modipafant (UK-74505) | 122956-68-7 | 98.15% | 605.09 | 500 MG
SDP
Supplier:  Medchemexpress LLC HY10890850MG
Encompass_Preferred
(Rac)-Modipafant (UK-74505) is an orally active, selective, and long-acting irreversible platelet activating factor receptor (PAFR) antagonist. It prevents dengue infection and is intended for research use only.
  • Orally active
  • Selective PAFR antagonist
  • Long-acting irreversible PAFR antagonist
  • Prevents dengue infection
  • For research use only
Catalog No. 50-003-46039