Learn More
Medchemexpress LLC (Rac)-Moxifloxacin | 354812-41-2 | 99.5% | C21H24FN3O4 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY66011B1MG
Encompass_Preferred
(Rac)-Moxifloxacin, also known as (Rac)-BAY 12-8039 free base, is an isoform of Moxifloxacin Hydrochloride. It functions as an oral 8-methoxyquinolone antimicrobial, primarily used in the treatment of various bacterial infections including acute bacterial sinusitis, acute bacterial exacerbations of chronic bronchitis, and community-acquired pneumonia.
  • Functions as an oral 8-methoxyquinolone antimicrobial
  • Used in treatment of acute bacterial sinusitis
  • Used in treatment of acute bacterial exacerbations of chronic bronchitis
  • Used in treatment of community-acquired pneumonia
  • Rapid action against Listeria monocytogenes
  • Demonstrates complete broth sterilization within 24 hours
  • Offers protection against macrophage lysis
  • Significantly prolonged survival in in vivo studies
  • Reduced bacterial overgrowth in spleens and lungs without toxicity
Catalog No. 50-004-33847