Learn More
Medchemexpress LLC (Rac)-NSC305787 | 785718-37-8 | 99.3% | 445.42 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY1893110MM/1ML
Encompass_Preferred
NSC305787 is an inhibitor of ezrin with a Kd of 5.85 μM. It inhibits the phosphorylation of ezrin caused by PKCΙ with an IC50 of 8.3 μM, and exhibits antitumor activity. NSC305787 inhibits PKCΙ phosphorylation of Ezrin, Moesin, Radixin, and MBP, with IC50s of 8.3, 9.4, 55, and 58.9 μM, respectively. It binds to PKCΙ with a Kd value of 172.4 μM, and inhibits ezrin T567 phosphorylation primarily by binding to ezrin, rather than through inhibition of PKCΙ kinase activity.
  • Shows inhibitory activity against ezrin-mediated invasion of K7M2 osteosarcoma cells.
  • Reduces cell motility phenotypes in zebrafish and blocks osteosarcoma metastatic growth in lung organ culture.
  • Suppresses ezrin-dependent osteosarcoma metastatic growth in mouse lung.
  • Inhibits pulmonary metastasis in a transgenic mouse model of osteosarcoma.
Catalog No. 50-003-85233