Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (Rac)-NSC305787 hydrochloride | 53868-26-1 | MFCD29924224 | 98.2% | 481.89 | C25H31Cl3N2O | 10 MG
SDP

Supplier:  Medchemexpress LLC HY18931A10MG

Encompass_Preferred

(Rac)-NSC305787 hydrochloride is a small-molecule ezrin inhibitor supplied as a hydrochloride salt for research use. It binds ezrin with micromolar affinity and inhibits ezrin phosphorylation, showing antitumor activity in preclinical studies.

  • Inhibits ezrin with Kd 5.85 μM.
  • Inhibits ezrin phosphorylation (IC50 8.3 μM).
  • High purity (98.18%).
  • Molecular weight 481.89 g/mol.
  • Solid, hygroscopic; soluble in DMSO (8 mg/mL), insoluble in water.
  • Recommended storage: solid at 4°C; solutions -80°C (6 months), -20°C (1 month).
  • Available in multiple pack sizes including 10 mg.

Catalog No. 50-002-94767


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.