Learn More
Medchemexpress LLC (Rac)-P1D-34 | 2957895-04-2 | 99.45% | 835.45 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1713345MG
Encompass_Preferred
(Rac)-P1D-34 is a PROTAC-based PIN1 degrader. It is composed of a PIN1 ligand, a cereblon ligand, and a linker. This product possesses anti-cancer activity, with IC50 values of 2248 nM in MV-4-11 cells, 3984 nM in MOLM-13 cells, and 3925 nM in HL-60 cells. This product is for research use only.
  • PROTAC-based PIN1 degrader
  • Composed of a PIN1 ligand, a cereblon ligand, and a linker
  • Exhibits anti-cancer activity in various cell lines
  • Suitable for research applications
Catalog No. 50-003-86240