Learn More
Medchemexpress LLC (Rac)-P1D-34 | 2957895-04-2 | 99.5% | 10 MG
SDP
Supplier:  Medchemexpress LLC HY17133410MG
Encompass_Preferred
(Rac)-P1D-34 is a racemic PROTAC degrader that induces degradation of the peptidyl-prolyl isomerase Pin1 in cellular models. It has been characterized in biochemical and cell-based studies showing Pin1 loss, downstream client protein modulation, antiproliferative activity in leukemia lines, and induction of apoptosis and DNA damage.
Catalog No. 50-002-95321