Learn More
Medchemexpress LLC (Rac)-P1D-34 | 2957895-04-2 | 99.5% | 835.45 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY171334
Encompass_Preferred
This PROTAC-based degrader targets PIN1 (peptidylprolyl cis/trans isomerase, NIMA-interacting 1). It is composed of a PIN1 ligand, a cereblon ligand, and a linker. This compound has demonstrated anti-cancer activity in various cell lines, including MV-4-11, MOLM-13, and HL-60 cells.
  • PROTAC-based degrader.
  • Targets PIN1.
  • Exhibits anti-cancer activity.
    • Effective in MV-4-11 cells with an IC50 of 2248 nM.
    • Effective in MOLM-13 cells with an IC50 of 3984 nM.
    • Effective in HL-60 cells with an IC50 of 3925 nM.
Catalog No. 50-003-85135