Learn More
Medchemexpress LLC (Rac)-PT2399 | 1672662-07-5 | 99.8% | 419.32 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY108697A100MG
Encompass_Preferred
(Rac)-PT2399 (Compound 10e) is the racemate of PT2399, functioning as a potent and specific hypoxia-inducible factor 2a (HIF-2α) inhibitor. It exhibits an IC50 of 0.01 μM, making it suitable for targeted research.
  • Potent and specific HIF-2α inhibitor
  • IC50 of 0.01 μM
  • High purity of 99.84%
  • Molecular weight of 419.32
  • Suitable for research in cancer and metabolic pathways
Catalog No. 50-003-46606