Learn More
Medchemexpress LLC (Rac)-PT2399 | 1672662-07-5 | 99.84% | 419.32 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY108697A10MM/1ML
Encompass_Preferred
(Rac)-PT2399 (Compound 10e), the racemate of PT2399, acts as a potent and specific hypoxia-inducible factor 2a (HIF-2α) inhibitor with an IC50 of 0.01 μM. This product is for research use only and is not sold to patients.
  • Potent and specific hypoxia-inducible factor 2a (HIF-2α) inhibitor
  • IC50: 0.01 μM (HIF-2α)
  • Appearance: solid
  • Color: white to off-white
Catalog No. 50-003-46320