Learn More
Medchemexpress LLC Rac-Rasagiline 100mg | 1875-50-9 | 171.24 g/mol | C12H13N | 100 MG
SDP
Supplier:  Medchemexpress LLC HY117173100MG
Encompass_Preferred
(Rac)-Rasagiline (AGN1135) is the racemic form of rasagiline, a selective monoamine oxidase B (MAO-B) inhibitor used in biochemical and pharmacological research. Supplied as a 100 mg solid, the compound has molecular formula C12H13N, molecular weight 171.24 g/mol, and reported purity ≥97%.
  • Racemic form for enantiomeric comparison studies.
  • Selective MAO-B inhibition for neuropharmacology research.
  • High purity suitable for analytical and preclinical assays.
  • Compact 100 mg sample convenient for small-scale experiments.
Catalog No. 50-002-26354