Learn More
Medchemexpress LLC Rac-Rasagiline 1g | 1875-50-9 | 171.24 | C12H13N | 1 G

Supplier: Medchemexpress LLC HY1171731G
(Rac)-Rasagiline is the racemic form of rasagiline, a selective monoamine oxidase B (MAO-B) inhibitor used in neuroscience and Parkinson's disease research. Supplied as a research-grade chemical, it is provided in multiple pack sizes and accompanied by datasheet and certificate of analysis documents for laboratory use.
- Selective MAO-B inhibitor for Parkinson's disease research.
- Racemic mixture appropriate for biochemical and cellular assays.
- Stated purity ≥97.0% for consistent experimental results.
- Available in milligram to gram pack sizes, including a 1 g option.
- Molecular formula C12H13N; molecular weight 171.24 g/mol.
- Physical appearance: light brown to yellow liquid.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.