Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Rac-Rasagiline 500mg | 1875-50-9 | 171.24 g/mol | C12H13N | 500 MG
SDP

Supplier:  Medchemexpress LLC HY117173500MG

Encompass_Preferred

Rac-rasagiline is the racemic form of rasagiline, a selective monoamine oxidase B (MAO-B) inhibitor used in preclinical Parkinson's disease and neuroprotection research. The compound is supplied as a light brown to yellow liquid with high purity for biochemical and pharmacological studies.

  • Selective MAO-B inhibitor suitable for Parkinson's disease models.
  • Demonstrated neuroprotective effects against MPTP-induced toxicity.
  • High purity (≥97.0%) for research use.
  • Molecular formula C12H13N and molecular weight 171.24 g/mol.
  • Store at low temperature for long-term stability (recommended -20°C).

Catalog No. 50-002-26356


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.