Learn More
Medchemexpress LLC (Rac)-Razpipadon | 1643462-93-4 | 97.1% | C19H17F2N3O4 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY141495
Encompass_Preferred
(Rac)-Razpipadon, also known as PW0464, is a nanomolar potent complete G protein biased ligand and a noncatechol D1R agonist. It exhibits agonist activity at D5R and wild-type human D1R in HEK293 cells and shows complete G protein bias without activity for D1R-mediated β-arrestin recruitment. This compound is intended for research use only.
  • Nanomolar potent complete G protein biased ligand
  • Noncatechol D1R agonist with EC50 of 5.8 nM (Gs-cAMP)
  • Agonist activity at D5R (EC50 12 nM) and human D1R (EC50 5.3 nM) in HEK293 cells
  • Exhibits complete G protein bias in vitro
  • Appears as a white to off-white solid
  • Soluble in DMSO at 25 mg/mL
Catalog No. 50-003-52597