Learn More
Medchemexpress LLC (Rac)-Razpipadon | 1643462-93-4 | 97.1% | C19H17F2N3O4 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY14149550MG
Encompass_Preferred
(Rac)-Razpipadon is a noncatechol D1R agonist that functions as a nanomolar potent complete G protein biased ligand, exhibiting an EC50 of 5.8 nM (Gs-cAMP).
  • Demonstrates complete G protein bias and does not show activity for D1R-mediated β-arrestin recruitment.
  • As a non-catechol agonist, forms bonds with S1985.42 and S2025.46 via its fluorine atom.
Catalog No. 50-003-53152