Learn More
Medchemexpress LLC (Rac)-Reparixin | 957407-64-6 | C14H21NO3S | 5 MG
SDP
Supplier:  Medchemexpress LLC HY15251A5MG
Encompass_Preferred
(Rac)-Reparixin (HY-15251A) is an isomer of Reparixin, used as an experimental control. It acts as a non-competitive allosteric inhibitor of the chemokine receptors CXCR1 and CXCR2 activation, with IC50s of 1 nM and 100 nM respectively. This compound targets CXCR and is applicable in research areas such as inflammation/immunology, endocrinology, and cancer.
  • Isomer of Reparixin
  • Used as an experimental control
  • Non-competitive allosteric inhibitor of chemokine receptors CXCR1 and CXCR2
  • Inhibits CXCR1 with an IC50 of 1 nM
  • Inhibits CXCR2 with an IC50 of 100 nM
  • Targets CXCR
  • Applicable in inflammation/immunology research
  • Applicable in endocrinology research
  • Applicable in cancer research
Catalog No. 50-003-72379