Learn More
Medchemexpress LLC (Rac)-Rhododendrol ((Rac)-betuligenol) | 69617-84-1 | 99.8% | 166.22 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY13691310MM/1ML
Encompass_Preferred
(Rac)-Rhododendrol ((Rac)-Betuligenol) is an aromatic compound recognized for its pro-oxidant activity. It has potential applications in suppressing liver diseases. However, it can also be toxic to melanocytes, inducing cell death.
  • Aromatic compound with pro-oxidant activity
  • May be useful in the suppression of liver diseases
  • Can be toxic to melanocytes, leading to cell death
  • Metabolite, RD-quinone, is cytotoxic and causes enzyme inactivation and endoplasmic reticulum stress by binding to thiol proteins
  • Derived melanin exhibits potent pro-oxidant activity and may cause oxidative stress
Catalog No. 50-003-48806