Learn More
Medchemexpress LLC (Rac)-Rhododendrol ((Rac)-betuligenol) | 69617-84-1 | 99.8% | 166.22 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY136913100MG
Encompass_Preferred
(Rac)-Rhododendrol, also known as (Rac)-Betuligenol, is an aromatic compound exhibiting pro-oxidant activity. It shows potential in suppressing liver diseases but can be toxic to melanocytes, leading to cell death. Its metabolite, RD-quinone, is cytotoxic and can cause enzyme inactivation and endoplasmic reticulum stress by binding to thiol proteins. The compound is intended for research use only.
  • Aromatic compound with pro-oxidant activity
  • May be beneficial in suppressing liver diseases
  • Can be toxic to melanocytes, leading to cell death
  • Metabolite RD-quinone is cytotoxic
  • Intended for research use only
Catalog No. 50-003-48491