Learn More
Medchemexpress LLC (Rac)-Rotigotine hydrochloride | 102120-99-0 | C19H26ClNOS | 100 MG
SDP
Supplier:  Medchemexpress LLC HY15394100MG
Encompass_Preferred
(Rac)-Rotigotine hydrochloride is a racemate of Rotigotine, functioning as a full agonist of the dopamine receptor, a partial agonist of the 5-HT1A receptor, and an antagonist of the α2B-adrenergic receptor. It exhibits high selectivity for the D3 receptor with a Ki of 0.71 nM and is intended for research use only.
  • Functions as a full agonist of the dopamine receptor
  • Partial agonist of the 5-HT1A receptor
  • Antagonist of the α2B-adrenergic receptor
  • Selective for D3 receptors over D2, D4, D5, and D1 receptors
  • Protects dopaminergic neurons against MPP+ toxicity and inhibits rotenone-induced ROS production
  • Soluble in DMSO (≥ 50 mg/mL)
Catalog No. 50-003-76702