Learn More
Medchemexpress LLC (Rac)-Rotigotine hydrochloride | 102120-99-0 | C19H26ClNOS | 50 MG
SDP
Supplier:  Medchemexpress LLC HY1539450MG
Encompass_Preferred
(Rac)-Rotigotine hydrochloride is a racemate of Rotigotine, functioning as a full agonist of the dopamine receptor, a partial agonist of the 5-HT1A receptor, and an antagonist of the α2B-adrenergic receptor. It exhibits high selectivity for D3 receptors and is intended for research use only.
  • Acts as a full agonist at all dopamine receptors
  • Functions as a partial agonist of the 5-HT1A receptor
  • Antagonizes the α2B-adrenergic receptor
  • Demonstrates 10-fold selectivity for D3 receptors over D2, D4, and D5
  • Provides significant protection against rotenone-induced cell death
  • Inhibits rotenone-induced ROS production
Catalog No. 50-003-76319