Learn More
Medchemexpress LLC (Rac)-SCH 563705 | 2701435-64-3 | 99.8% | 425.48 g/mol | C23H27N3O5 | 100 MG

Supplier: Medchemexpress LLC HY10011B100MG
This racemic small-molecule antagonist of CXCR1 and CXCR2 is used in research on chemokine signaling and inflammatory responses. It demonstrates nanomolar potency and is supplied as a high-purity solid suitable for in vitro pharmacology and in vivo studies.
- Potent CXCR1 and CXCR2 antagonist with reported IC50 values in the low nanomolar range.
- High purity (≈99.8%) for reliable experimental consistency.
- Molecular weight 425.48 g/mol for stoichiometric calculations.
- Available in multiple pack sizes, including 100 MG, for flexible study scales.
- CAS number 2701435-64-3 provided for unambiguous identification.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.