Learn More
Medchemexpress LLC (Rac)-Shikonin | 54952-43-1 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYN60045MG
Encompass_Preferred
(Rac)-Shikonin, also known as Shikalkin, is a compound that exhibits anti-tumor activity. It is notable for its ability to circumvent cancer agent resistance by inducing a necroptotic death pathway. This compound has shown similar potency against drug-sensitive cancer cell lines, such as MCF-7 and HEK293, as well as their drug-resistant counterparts that overexpress P-glycoprotein, Bcl-2, or Bcl-xL.
  • Possesses anti-tumor activity
  • Circumvents cancer agent resistance by induction of a necroptotic death
  • Shows similar potency against drug-sensitive and drug-resistant cancer cell lines
  • Should be stored as a solid at 4°C, protected from light
  • In solvent, store at -80°C for 6 months or -20°C for 1 month, protected from light
Catalog No. 50-004-21660