Learn More
Medchemexpress LLC (rac)-shikonin | 54952-43-1 | MFCD00016668 | 99.0% | 288.30 g/mol | C16H16O5 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYN600410MG
Encompass_Preferred
(Rac)-Shikonin is the racemic form of shikonin, a naphthoquinone natural product used in research to induce necroptosis and investigate cell-death pathways. It is provided as a high-purity solid suitable for in vitro laboratory studies.
  • Induces necroptotic and necrotic cell death in cancer model systems.
  • Useful for studying cell-death pathways and mechanisms of drug resistance.
  • High purity solid appropriate for research applications.
  • Stable when stored protected from light; specific solvent storage conditions apply.
  • Available in small (mg-scale) quantities for laboratory use.
Catalog No. 50-002-89701