Learn More
Medchemexpress LLC (Rac)-sn-Glycerol 3-phosphate sodium | 1555-56-2 | 99.95% | 216.04 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY108776100MG
Encompass_Preferred
(Rac)-sn-Glycerol 3-phosphate sodium is an α-site substrate analogue that strongly inhibits the reaction rate of indole and nucleophilic indole analogues with E(A-A) when bound to the α-site. It is a phosphorylated derivative of glycerol produced as an intermediate in the glycolysis pathway. It also binds to the α subunit of tryptophan synthase, regulating tryptophan synthesis through allosteric inhibition.
  • Provides microorganisms with phosphorus, carbon, and energy sources in nutrient-lacking environments.
  • Usable for microbial cultivation.
  • Binds to metal ions, forming insoluble phosphate minerals.
  • Can be used for in situ bioremediation to reduce environmental pollution.
Catalog No. 50-003-46246