Learn More
Medchemexpress LLC 1,2,3-Propanetriol, 1-(dihydrogen phosphate), sodium salt (1:2) | 1555-56-2 | 100.0% | 216.04 | 250 MG
Supplier: Medchemexpress LLC 250MGHY108776
(Rac)-sn-Glycerol 3-phosphate sodium is an α-site substrate analogue that strongly inhibits the reaction rate of indole and nucleophilic indole analogues with E(A-A). It is a phosphorylated derivative of glycerol, produced as an intermediate in the glycolysis pathway, regulating tryptophan synthesis through allosteric inhibition. This compound can serve as a phosphorus, carbon, and energy source for microorganisms, making it suitable for microbial cultivation. It also binds to metal ions, forming insoluble phosphate minerals, which can be used for in situ bioremediation.
- Endogenous metabolite
- Alpha-site substrate analogue
- Regulates tryptophan synthesis
- Can be used for microorganisms cultivation
- Can be used for in situ bioremediation