Learn More
Medchemexpress LLC (Rac)-Sograzepide | 168161-71-5 | 498.58 | 1 ML
SDP
Supplier:  Medchemexpress LLC HYU0036010MM/1ML
Encompass_Preferred
(Rac)-Sograzepide is an antagonist of cholecystokinin B (CCK-B) receptor, and has the potential of reducing the secretion of gastric acid. Storage recommendations are -20°C for 3 years (powder) or 4°C for 2 years (powder), and -80°C for 2 years (in solvent) or -20°C for 1 year (in solvent).
  • Antagonist of cholecystokinin B (CCK-B) receptor
  • Reduces secretion of gastric acid
  • Targets cholecystokinin receptor
  • Involves GPCR/G protein and neuronal signaling pathways
Catalog No. 50-004-47119