Learn More
Medchemexpress LLC (Rac)-Sograzepide | 168161-71-5 | 498.58 | 50 MG
SDP
Supplier:  Medchemexpress LLC HYU0036050MG
Encompass_Preferred
(Rac)-Sograzepide is an antagonist of cholecystokinin B (CCK-B) receptor, and has the potential of reducing the secretion of gastric acid. It is an extremely potent, highly selective, and orally active Gastrin/CCK-B antagonist.
  • Antagonist of cholecystokinin B (CCK-B) receptor
  • Potential for reducing secretion of gastric acid
  • Solubility: DMSO : 125 mg/mL (ultrasonic)
  • No development reported in clinical information
  • Shipping at room temperature
Catalog No. 50-004-49512