Learn More
Medchemexpress LLC Rac-Sphingosylphosphorylcholine | 82970-80-7 | >98% | 464.62 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1651355MG
Encompass_Preferred
Rac-Sphingosylphosphorylcholine is a phospholipid-derived compound characterized by a long alkyl chain backbone, an amino group at C2, a hydroxyl group at C3, and an (E)-configured double bond at C2, plus a 2-(trimethylazaniumyl)ethyl phosphate group. It is synthesized as a high-purity pharmaceutical intermediate, often used in studies of lipid metabolism and membrane biology.
  • Plays a critical role in cellular processes like proliferation, differentiation, and apoptosis.
  • Acts as a signaling molecule, interacting with G-protein coupled receptors.
  • May offer neuroprotective effects, promoting neuronal survival in neurological disorders.
  • Influences vascular smooth muscle cells and endothelial function.
  • Explored for its effects on tumor growth and metastasis.
  • Modulates immune responses, affecting innate and adaptive immunity.
  • Integral to cell membrane formation, stability, fluidity, and integrity.
Catalog No. 50-003-91782