Learn More
Medchemexpress LLC (rac)-TBAJ-5307 | 2131783-85-0 | 95% | 627.53 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY161135A5MG
Encompass_Preferred
(rac)-TBAJ-5307 is the racemate of TBAJ-5307, a broad spectrum anti-non-tuberculous mycobacteria inhibitor that can target the FO-domain of the engine and prevent rotation and proton-translocation.
  • Targets bacterial pathogens
  • Reference compound
Catalog No. 50-003-93170