Learn More
Medchemexpress LLC (Rac)-TBAJ-876 | 2131784-39-7 | 99.98% | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY128866A
Encompass_Preferred
(Rac)-TBAJ-876 is a racemate of TBAJ-876, which acts as an inhibitor of *Mycobacterium tuberculosis*. This compound is an analogue of the anti-tuberculosis agent Bedaquiline (HY-14881) and shows potential for tuberculosis research.
  • Racemate of TBAJ-876
  • Inhibits *Mycobacterium tuberculosis*
  • Analogue of Bedaquiline (HY-14881)
  • Suitable for tuberculosis research
Catalog No. 50-003-42946