Learn More
Medchemexpress LLC (Rac)-telmesteine | 127657-29-8 | 98.0% | 205.24 | C7H11NO4S | 1 G

Supplier: Medchemexpress LLC HY1082851G
Telmesteine (racemic) is a small-molecule protease inhibitor used as an enzyme stabilizer in protease-containing detergents and cleaning agents. It is supplied for research use in solid and solution formats and is accompanied by safety and analytical documentation.
- Protease inhibitor and enzyme stabilizer.
- Provided as solid and as 10 mM solution in DMSO.
- High purity: 98.0%.
- Molecular weight: 205.24 g/mol.
- Recommended storage: powder -20°C (long term) or 4°C; in solvent -80°C or -20°C.
- Documentation includes SDS, COA, and data sheet.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.