Learn More
Medchemexpress LLC (Rac)-Tenofovir alafenamide-d5 | 379270-36-7 | 481.50 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYW004356S1MG
Encompass_Preferred
(2R)-Isopropyl 2-((((((R)-1-(6-amino-9H-purin-9-yl)propan-2-yl)oxy)methyl)(phenoxy)phosphoryl)amino)propanoate-d5 is a deuterium labeled compound. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process. Deuteration has gained attention due to its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer.
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-48822